Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK R (CD266) Mouse, anti-Human, R718, Clone: ITEM-4, BD OptiBuild™

Catalog No. BDB751739 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751739 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751739 Supplier BD Biosciences Supplier No. 751739
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ITEM-4 monoclonal antibody specifically binds to the TWEAK Receptor (TWEAK-R/TWEAKR), which is also known as CD266, or Fibroblast growth factor inducible 14 (FGF-inducible 14, FN14). The TWEAK Receptor is a ∼14 kDa type I transmembrane protein that is the 12A family member of the tumor necrosis factor receptor superfamily (TNFRSF12A). It is expressed at low levels in a variety of tissues including, heart, placenta, lung, muscle, and pancreas. TWEAK Receptors are relatively highly expressed on HUVEC cells and certain tumor cell lines. TWEAK-induced TWEAK Receptor-mediated signaling can play roles in inflammation, the induction of apoptosis in certain cell types, the proliferation and migration of endothelial cells, and can promote angiogenesis within healthy and diseased tissues, eg, tumors. The ITEM-4 antibody can induce the death of certain TWEAK-sensitive tumor target cell lines. The TWEAK Receptor binds TWEAK (CD255) and can signal intracellularly by interactions with TRAF1, 2, and 3 cytoplasmic proteins leading to NF-κB activation. The ITEM-4 antibody reportedly crossreacts strongly with mouse CD266 as demonstrated by flow cytometric analysis and functional assays.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TWEAK R (CD266)
Applications Flow Cytometry
Classification Monoclonal
Clone ITEM-4
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.