Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TWEAK R/TNFRSF12 Antibody (314636) [Allophycocyanin], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB16101A
Description
TWEAK R/TNFRSF12 Monoclonal antibody specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
TWEAK R/TNFRSF12 | |
Monoclonal | |
APC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
314636 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
E. coli-derived recombinant mouse TWEAK R, Glu28-Trp79, Accession # Q9CR75.1 | |
0.1 mL | |
Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors | |
51330 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction