Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TWEAK R/TNFRSF12 Antibody (ITEM-4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200409AF750
Description
TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
TWEAK R/TNFRSF12 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TNFRSF12A | |
The immunogen for this antibody is TWEAKR. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
ITEM-4 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
Mouse | |
Protein A or G purified | |
Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors | |
51330.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction