Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK R/TNFRSF12 Antibody (ITEM-4), Biotin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$304.50 - $646.00

Specifications

Antigen TWEAK R/TNFRSF12
Clone ITEM-4
Dilution Flow Cytometry 1:10-1:1000
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14348303
SDP
View Documents
Novus Biologicals
NBP1434830.025MG
0.025 mg
Each for $304.50
Only null left
Add to Cart
 
NBP14348301
SDP
View Documents
Novus Biologicals
NBP1434830.1MG
0.1 mg
Each for $646.00
Only null left
Add to Cart
 
Description

Description

TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

TWEAK R/TNFRSF12
Flow Cytometry 1:10-1:1000
Monoclonal
Mouse
Angiogenesis, Apoptosis, Cancer, Tumor Suppressors
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
51330
The immunogen for this antibody is TWEAKR.
Primary
Store at 4C. Do not freeze.
ITEM-4
Flow Cytometry
Biotin
RUO
Human, Mouse
CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14
TNFRSF12A
IgG2b κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.