Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK Receptor (CD266) Mouse, Brilliant Violet 605, Clone: ITEM-4, BD Optibuild™

Catalog No. BDB742692 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742692 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742692 Supplier BD Biosciences Supplier No. 742692
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ITEM-4 monoclonal antibody specifically binds to the TWEAK Receptor (TWEAK-R/TWEAKR), which is also known as CD266, or Fibroblast growth factor inducible 14 (FGF-inducible 14, FN14). The TWEAK Receptor is a ñ14 kDa type I transmembrane protein that is the 12A family member of the tumor necrosis factor receptor superfamily (TNFRSF12A). It is expressed at low levels in a variety of tissues including, heart, placenta, lung, muscle, and pancreas. TWEAK Receptors are relatively highly expressed on HUVEC cells and certain tumor cell lines. TWEAK-induced TWEAK Receptor-mediated signaling can play roles in inflammation, the induction of apoptosis in certain cell types, the proliferation and migration of endothelial cells, and can promote angiogenesis within healthy and diseased tissues, eg, tumors. The ITEM-4 antibody can induce the death of certain TWEAK-sensitive tumor target cell lines. The TWEAK Receptor binds TWEAK (CD255) and can signal intracellularly by interactions with TRAF1, 2, and 3 cytoplasmic proteins leading toNF-κBactivation. The ITEM-4 antibody reportedly crossreacts strongly with mouse CD266 as demonstrated by flow cytometric analysis and functional assays.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWEAK Receptor (CD266)
Classification Monoclonal
Clone ITEM-4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TWEAKR, TWEAK-R, TWEAK Receptor, TNFRSF12A, FGF-inducible 14, FN14
Host Species Mouse
Immunogen Human Fn14 transfected P815
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.