Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK Receptor (CD266) Mouse, Brilliant Violet 650, Clone: ITEM-4, BD Optibuild™

Catalog No. BDB742693 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742693 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742693 Supplier BD Biosciences Supplier No. 742693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ITEM-4 monoclonal antibody specifically binds to the TWEAK Receptor (TWEAK-R/TWEAKR), which is also known as CD266, or Fibroblast growth factor inducible 14 (FGF-inducible 14, FN14). The TWEAK Receptor is a ñ14 kDa type I transmembrane protein that is the 12A family member of the tumor necrosis factor receptor superfamily (TNFRSF12A). It is expressed at low levels in a variety of tissues including, heart, placenta, lung, muscle, and pancreas. TWEAK Receptors are relatively highly expressed on HUVEC cells and certain tumor cell lines. TWEAK-induced TWEAK Receptor-mediated signaling can play roles in inflammation, the induction of apoptosis in certain cell types, the proliferation and migration of endothelial cells, and can promote angiogenesis within healthy and diseased tissues, eg, tumors. The ITEM-4 antibody can induce the death of certain TWEAK-sensitive tumor target cell lines. The TWEAK Receptor binds TWEAK (CD255) and can signal intracellularly by interactions with TRAF1, 2, and 3 cytoplasmic proteins leading toNF-κBactivation. The ITEM-4 antibody reportedly crossreacts strongly with mouse CD266 as demonstrated by flow cytometric analysis and functional assays.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWEAK Receptor (CD266)
Classification Monoclonal
Clone ITEM-4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TWEAKR, TWEAK-R, TWEAK Receptor, TNFRSF12A, FGF-inducible 14, FN14
Host Species Mouse
Immunogen Human Fn14 transfected P815
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.