Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tween™ 20 Surfact-Amps™ Detergent Solution, 6 x 10 mL PROMO

Catalog No. PI28320PM
Click to view available options
:
Thermo Scientific™ Tween™ 20 Surfact-Amps™ Detergent Solution

Thermo Scientific™ Surfact-Amps 20 Detergent Solution is highly-purified Tween-20 detergent stabilized and supplied as a 10% solution in 10 mL glass ampules.

This is an easy-to-use 10% (w/v) solution of purified Tween-20 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Tween-20 is a nonionic polyoxyethylene surfactant that is most frequently added to PBS or TBS wash buffers for ELISA, Western blotting and other immunoassay methods.

Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

Tween 20 – a popular nonionic detergent for use in immunoassay wash buffers

Accurate – precise 10% detergent solution in ultrapure water

Easy-to-use – solution is simple to dispense and dilute for use

Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

Stable – packed in glass ampules under inert nitrogen gas

Detergent Properties:

  • Molecular Weight: 1228
  • Detergent Class: Nonionic
  • Aggregation Number: Unknown
  • Micelle Molecular Weight: Unknown
  • Critical Micelle Concentration (CMC): 0.06mM (0.0074%, w/v)
  • Cloud Point: 95°C
  • Dialyzable: No

;

Specifications

Product Type Detergent
Content And Storage Store in a cool, dry, and well-ventilated area
Form Liquid
Product Line Surfact-Amps™, Tween™
Reagent Type Detergent Solution
Quantity 6 x 10 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.