Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 60

Catalog No. p-3776705
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 9005-67-8 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | 522.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-67-8
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD01780110
InChI Key DEKFZWMENCCOFU-UHFFFAOYNA-N
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Acid Value 2mg KOH/g max.
Physical Form Paste at 25°C
Density 1.0800g/mL
Quantity 250 mL
Flash Point >150°C
Hydroxyl Value 81 to 96mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Saponification Value 45 to 55mg KOH/g
Specific Gravity 1.08
Viscosity 75-175 Pa.s (50°C)
Water 3% max.
Solubility Information Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, xylene, insoluble in propylene glycol and mineral oil
Chemical Name or Material Polysorbate 60
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.