Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Tween™ 80, MP Biomedicals™

Catalog No. mp021947251 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021947251 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021947251 Supplier MP Biomedicals™ Supplier No. 02194725.1
Only null left
Add to Cart
Add to Cart

Tween™ 80 is widely used in a number of biochemical applications.

Tweens™ are a series of nonionic surfactants derived from sorbitan esters.

  • They are soluble or dispersible in water but differ widely in organic and oil solubilities.
  • Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc.
  • Polysorbate 20 is nonionic surfactants derived from sorbitan ester.
TRUSTED_SUSTAINABILITY

Specifications

Description Tween™ 80
CAS 9005-65-6
Synonym Polyoxyethylenesorbitan monooleate; POE (20) sorbitan monooleate; Polyethylene glycol sorbitan monooleate; Polysorbate 80
Content And Storage Store at Room Temperature
Form Liquid
For Use With (Application) Selective protein extraction; Isolation of nuclei from mammalian cell lines
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.