Learn More
Thermo Scientific™ Tween™ 80 Surfact-Amps™ Detergent Solution, 50 mL PROMO
Description
Thermo Scientific™ Surfact-Amps 80 Detergent Solution is highly-purified Tween-80 detergent stabilized and supplied as a 10% solution in 10 mL glass ampules.
This is an easy-to-use 10% (w/v) solution of purified Tween-80 detergent for use in routine and high-demand protein research methods and molecular biology techniques. Tween-80 is a nonionic polyoxyethylene surfactant that is most frequently added to PBS or TBS wash buffers for ELISA, Western blotting and other immunoassay methods.
Surfact-Amps™ Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Highlights:
Tween 80;– a nonionic detergent for use in wash buffers
Accurate;– precise 10% detergent solution in ultrapure water
Easy-to-use;– solution is simple to dispense and dilute for use
Exceptionally pure;– less than 1.0μeq/ml peroxides and carbonyls
Stable;– packed in glass ampules under inert nitrogen gas
Detergent Properties:
- Molecular Weight: 1310
- Detergent Class: Nonionic
- Aggregation Number: 60
- Micelle Molecular Weight: 76,000
- Critical Micelle Concentration (CMC): 0.012mM (0.0016%, w/v)
- Cloud Point: Unknown
- Dialyzable: No
Specifications
Specifications
| Product Type | Detergent |
| Content And Storage | Store in a cool, dry, and well-ventilated area |
| Form | Liquid |
| Product Line | Surfact-Amps, Tween |
| Reagent Type | Detergent Solution |
| Quantity | 50 mL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.