Learn More
TWF1 Mouse anti-Human, Clone: OTI10E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808882

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.Specifications
TWF1 | |
Monoclonal | |
Unconjugated | |
TWF1 | |
A6, PTK9 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5756 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI10E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q12792 | |
TWF1 | |
Human recombit protein fragment corresponding to amino acids 140-384 of human TWF1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.