Learn More
TWF1/PTK9 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305278AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis and Bovine
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.Specifications
TWF1/PTK9 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q12792 | |
TWF1 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TWF1 | |
A6, PTK9, A6 protein tyrosine kinase, PTK9 protein tyrosine kinase 9, protein A6, protein tyrosine kinase 9, twinfilin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5756 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.