Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWF2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA589341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589341 Supplier Thermo Scientific Supplier No. PA589341

Rabbit Polyclonal Antibody

Immunogen sequence: MAHQTGIHAT EELKEFFAKA RAGSVRLIKV VIEDEQLVLG ASQEPVGRWD QDYDRAVLPL LDAQQPCYLL YRLDSQNAQG FEWLFLAWSP DNSPVRLKML YAATRATVKK EFGGGHIKDE LFGTVKDDLS FAGYQKHLSS CAAPAPLTSA ERELQQIRIN EVKTEISVES KHQTLQGLAF PLQPEAQRAL QQLKQKMVNY IQMKLDLERE TIELVHTEPT DVAQLPSRVP RDAARYHFFL YKHTHEGDPL ESVVFIYSMP GYKCSIKERM LYSSCKSRLL DSVEQDFHLE IAKKIEIGDG AELTAEFLYD EVHPKQHAFK QAFAKPKGPG GKRGHKRLIR GPGENGDDS; Positive Samples: Jurkat, 293T, A-431, Mouse heart, Mouse thymus; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, perinuclear region, stereocilium

TWF2 encodes a protein that was identified by its interaction with the catalytic domain of protein kinase C-zeta. The encoded protein contains an actin-binding site and an ATP-binding site. It is most closely related to twinfilin, a conserved actin monomer-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWF2
Applications Immunoprecipitation, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3
Gene TWF2
Gene Accession No. Q6IBS0, Q9Z0P5
Gene Alias A6 related protein, A6r, A6-related, A6-related protein, A6RP, AU014993, hA6RP, mA6RP, MSTP011, protein tyrosine kinase 9 related protein, Protein tyrosine kinase 9-like, protein tyrosine kinase 9-like (A6-related protein), PTK 9L, PTK9 protein typrotein tyrosine kinase 9-like (A6-related protein), Ptk9l, PTK9L protein tyrosine kinase 9-like (A6-related protein), Ptk9r, TWF 2, Twf2, Twinfilin 2, twinfilin actin binding protein 2, twinfilin actin-binding protein 2, twinfilin, actin-binding protein, homolog 2, twinfilin, actin-binding protein, homolog 2 (Drosophila), twinfilin-1-like protein, Twinfilin2, Twinfilin-2
Gene Symbols TWF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human TWF2 (NP_009215.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11344, 23999, 684352
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.