Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Twist-2 Antibody (1A11D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Twist-2 |
---|---|
Clone | 1A11D9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry |
Description
Twist-2 Monoclonal specifically detects Twist-2 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.Specifications
Twist-2 | |
1 mg/mL | |
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Inhibitors Activators and Regulators | |
PBS with 0.05% Sodium Azide | |
117581 | |
Purified recombinant fragment of human TWIST2 (AA: 1-160) expressed in E. Coli. | |
Primary | |
18.1 kDa |
1A11D9 | |
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BHLHA39, bHLHa39twist-related bHLH protein Dermo1, Class A basic helix-loop-helix protein 39, Dermis-expressed protein 1, Dermo-1, DERMO1MGC117334, twist homolog 2 (Drosophila), twist-related protein 2 | |
TWIST2 | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title