Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TWIST1/2 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 630770

Description
Positive control: SKBR3 Cellular location: Nucleus.
TWIST1/2 Polyclonal antibody specifically detects TWIST1/2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
TWIST1/2 | |
Polyclonal | |
Unconjugated | |
TWIST1 | |
AA960487; ACS3; AMS; BBRSAY; B-HLH DNA binding protein; bHLHa38; BHLHA39; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; Class A basic helix-loop-helix protein 39; CRS; CRS1; CSO; dermis expressed 1; dermis-expressed protein 1; DERMO1; Dermo-1; dermo-1 protein; FFDD3; H-twist; MGC117334; M-Twist; Pde; pdt; Pluridigite; polydactyly EMS; SCS; SETLSS; Ska<m10Jus>; Ska<sup>m10Jus</sup>; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist basic helix-loop-helix transcription factor 2; twist family bHLH transcription factor 1; twist family bHLH transcription factor 2; twist gene homolog 1; twist homolog 1; twist homolog 2; TWIST homolog of drosophila; Twist1; Twist2; twist-related bHLH protein Dermo1; twist-related protein 1; Twist-related protein 2 | |
A synthetic peptide RL SYA FSV WRM EGA WSM SAS H corresponding to human TWIST protein | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P26687, P97831, Q15672, Q8WVJ9, Q9D030 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117581, 13345, 22160, 59327, 7291, 85489 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction