Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TWIST1 Recombinant Rabbit Monoclonal Antibody (BLR156J)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544498
This item is not returnable.
View return policy
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
TWIST1 acts as a transcriptional regulator. It inhibits myogensis by sequestering E proteins, inhibiting trans-activation by MEF2, and inhibiting DNA-binding by MYOD1 through physical interaction. TWIST1 also represses proinflammatory cytokines such as TNFA and IL1B. TWIST also regulates cranial suture patterning and fusion and gene expression differentially depending on dimer composition. Mutations in the gene are implicated in Saethre-Chotzen syndrome, a disease characterized by coronal synostosis, brachycephaly, low front hairline, facial asymmetry, hypertelorism, broad halluces, and clinodactyly.
Specifications
TWIST1 | |
Recombinant Monoclonal | |
0.25 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8-7.4 | |
Q15672 | |
TWIST1 | |
Peptide representing a region between residue 1 and 50 of Human Twist-related protein 1. | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
BLR156J | |
Unconjugated | |
TWIST1 | |
AA960487; ACS3; B-HLH DNA binding protein; bHLHa38; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; CRS; CRS1; CSO; H-twist; M-twist; p ACS3; Pde; pdt; Pluridigite; polydactyly EMS; SCS; Ska>m10Jus>; Ska>sup>m10Jus>/sup>; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist family bHLH transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist1; twist-related protein 1 | |
Rabbit | |
Purified | |
RUO | |
7291 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction