Learn More
Invitrogen™ Twist1/Twist2 Polyclonal Antibody

Description
Positive Control: Raji, THP-1, HL-60, TWIST1 transfected 293T lysates, human TWIST1-transfected 293T, Jurkat, OECM1, CL1-5, Toledo, SU-DHL-5, HT, DB, RL95-2, MDA-MB-231 Predicted Reactivity: Japanese Medaka (95%), Xenopus laevis (97%), Chicken (100%), Chimpanzee (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Twist1/Twist2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.67 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | TWIST1 |
| Gene Accession No. | P26687, P97831, Q15672, Q8WVJ9, Q9D030 |
| Gene Alias | AA960487; ACS3; AMS; BBRSAY; B-HLH DNA binding protein; bHLHa38; BHLHA39; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; Class A basic helix-loop-helix protein 39; CRS; CRS1; CSO; dermis expressed 1; dermis-expressed protein 1; DERMO1; Dermo-1; dermo-1 protein; FFDD3; H-twist; MGC117334; M-Twist; Pde; pdt; Pluridigite; polydactyly EMS; SCS; SETLSS; Ska>m10Jus>; Ska>sup>m10Jus>/sup>; Ska10; Twist; twist basic helix-loop-helix transcription factor 1; twist basic helix-loop-helix transcription factor 2; twist family bHLH transcription factor 1; twist family bHLH transcription factor 2; twist gene homolog 1; twist homolog 1; twist homolog 2; TWIST homolog of drosophila; Twist1; Twist2; twist-related bHLH protein Dermo1; twist-related protein 1; Twist-related protein 2 |
| Gene Symbols | TWIST1, TWIST2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.