Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TWIST2 Monoclonal Antibody (1A11D9)
GREENER_CHOICE

Catalog No. PIMA531944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531944 Supplier Invitrogen™ Supplier No. MA531944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31944 has been tested in indirect ELISA.

Binds to the E-box consensus sequence 5'-CANNTG-3' as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWIST2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A11D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene TWIST2
Gene Accession No. Q8WVJ9
Gene Alias AMS; BBRSAY; bHLHa39; Class A basic helix-loop-helix protein 39; dermis expressed 1; dermis-expressed protein 1; DERMO1; Dermo-1; dermo-1 protein; FFDD3; SETLSS; twist basic helix-loop-helix transcription factor 2; twist family bHLH transcription factor 2; twist homolog 2; TWIST2; twist-related bHLH protein Dermo1; twist-related protein 2
Gene Symbols TWIST2
Host Species Mouse
Immunogen Purified recombinant fragment of human TWIST2 (AA: 1-160) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117581
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.