Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TWIST2 Monoclonal Antibody (OTI6G9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA805300

Description
Binds to the E-box consensus sequence 5′-CANNTG-3′ as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.Specifications
TWIST2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WVJ9 | |
TWIST2 | |
Full length human recombit protein of human TWIST2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI6G9 | |
Unconjugated | |
TWIST2 | |
AMS, BBRSAY, DERMO1, FFDD3, SETLSS, bHLHa39 | |
Mouse | |
Affinity Chromatography | |
RUO | |
117581 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction