Learn More
TWIST2 Mouse anti-Human, Clone: OTI6H10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805212

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Binds to the E-box consensus sequence 5′-CANNTG-3′ as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.Specifications
TWIST2 | |
Monoclonal | |
Unconjugated | |
TWIST2 | |
AMS, BBRSAY, DERMO1, FFDD3, SETLSS, bHLHa39 | |
Mouse | |
Affinity Chromatography | |
RUO | |
117581 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8WVJ9 | |
TWIST2 | |
Full length human recombit protein of human TWIST2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.