Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TWIST2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TWIST2 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7427
![]() ![]() |
Proteintech Group Inc
117521AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Binds to the E-box consensus sequence 5′-CANNTG-3′ as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.Specifications
TWIST2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8WVJ9, Q9D030 | |
117581, 13345 | |
TWIST2 Fusion Protein Ag2348 | |
Primary | |
-20°C | |
TWIST2 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
bHLHa39, Dermis expressed protein 1, Dermo 1, DERMO1, twist homolog 2 (Drosophila), Twist related protein 2, TWIST2 | |
Twist2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title