Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Twistit: Four Stoppers in One
SDPSCI_ED

Multi-purpose and economical

Supplier:  Plasticoid H5M669

 View more versions of this product

Catalog No. S3508B


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Four stoppers in one! Seals twist off to adapt the stopper to your application. Use as a solid, one-, two-, or three-hole stopper. Reduces inventory, confusion, and cost. Made of clean, white, resilient, natural rubber with a clinical finish.

Specifications

Specifications

Twistit Stopper
2.7 cm
2.3 cm
Clinical finish
White
5
5
3
Multi-purpose applications
Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.