Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TWISTNB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554586 Supplier Invitrogen™ Supplier No. PA554586
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QTMEINMGDE LEFEVFRLDS DAAGVFCIRG KLNITSLQFK RSEVSEEVTE NGTEEAAKKP KKKKKKKDPE TYEVDSGTTK LAD Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.

DNA-directed RNA polymerase I subunit RPA43 or Twist neighbor protein (Twistnb)belongs to the eukaryotic RPA43 RNA polymerase subunit family. It catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates.Twistnb is the component of RNA polymerase I which synthesizes ribosomal RNA precursors. It is widely expressed in all fetal and adult tissues, with highest expression in fetal lung, liver, and kidney, and low expression in all adult tissues. Twist genes are essential for embryonic development and are conserved from jellyfish to human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWISTNB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TWISTNB
Gene Accession No. Q3B726
Gene Alias 2410173G11; 2810024J17Rik; D16Wsu83e; DNA-directed RNA polymerase I subunit RPA43; POLR1F; RNA polymerase I subunit F; twist basic helix-loop-helix transcription factor 1 neighbor; TWIST neighbor; twist neighbor protein; TWISTNB
Gene Symbols TWISTNB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TWISTNB. Recombinant protein control fragment (Product #RP-92636).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221830
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.