Learn More
Invitrogen™ TWISTNB Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 100% homologous in chimpanzee, and 94% homologous in rat and dog.
Specifications
Specifications
| Antigen | TWISTNB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | TWISTNB |
| Gene Accession No. | Q3B726, Q78WZ7 |
| Gene Alias | 2410173G11; 2810024J17Rik; D16Wsu83e; DNA-directed RNA polymerase I subunit RPA43; POLR1F; RNA polymerase I subunit F; twist basic helix-loop-helix transcription factor 1 neighbor; TWIST neighbor; twist neighbor protein; TWISTNB |
| Gene Symbols | TWISTNB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.