Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXK Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145195 Supplier Invitrogen™ Supplier No. PA5145195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSSYNTIQSV FCCCCCCSVQ KRQMRTQISL STDEELPEKY TQRRRPWLSQ LSNKKQSNTG RVQPSKRKPL PPLPPSEVAE EKIQVKALYD FLP Antigen sequence identity: Mouse 72%; Rat 63%.

TXK, along with the close relatives BMS, ITK, and BTK, signal downstream of antigen receptors and other types of receptors. All members of the family contain SH3 and SH2 domains and, with the exception of Txk and Dsrc28C, also contain a pleckstrin homology (PH) and a Tec homology (TH) domain in their amino termini. Four alternatively spliced forms of Tec are found to be expressed broadly in cells of hematopoietic lineage and hepatocytes. The Emt gene product associates with CD28 and becomes activated subsequent to CD28 ligation. Btk is necessary for proper B cell development, and mutations in the gene encoding Btk have been associated with families suffering from X-linked agammaglobulinemia, also referred to as Bruton's disease. The Bmx protein shares a high degree of homology with Btk and seems to be expressed at highest levels in the heart. Txk expression is T cell-specific, while expression of the Drosophila Tec homolog, Dsrc28C, is developmentally regulated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TXK
Gene Accession No. P42681
Gene Alias A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK
Gene Symbols TXK
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 3-95.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.