Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXK Polyclonal Antibody
TXK Polyclonal Antibody
Supplier: Thermo Scientific PA590043
Description
TXK Polyclonal Antibody for Western Blot

Specifications
TXK | |
Polyclonal | |
Unconjugated | |
TXK | |
TXK | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human TXK (NP_0033192). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
1.4 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
A130089B16Rik; BTKL; Protein-tyrosine kinase 4; PSCTK5; PTK4; PTK4 protein tyrosine kinase 4; PTK-RL-18; resting lymphocyte kinase; Rlk; TKL; TXK; TXK tyrosine kinase; tyrosine kinase; tyrosine-protein kinase TXK | |
Rabbit | |
Affinity Chromatography | |
RUO | |
305311 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction