Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXLNA Polyclonal Antibody

Catalog No. PIPA5121514
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121514 20 μL
PIPA5119453 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121514 Supplier Invitrogen™ Supplier No. PA5121514
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TXLNA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

May be involved in intracellular vesicle traffic and potentially in calcium-dependent exocytosis in neuroendocrine cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXLNA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TXLNA
Gene Accession No. P40222
Gene Alias Alpha-taxilin; DKFZp451J0118; il 14; IL14; IL-14; interleukin 14; MGC118870; MGC118871; RP4-622L5.4; taxilin alpha; TXLN; TXLNA
Gene Symbols TXLNA
Host Species Rabbit
Immunogen Recombinant protein Taxilin Alpha. The antigen corresponds to amino acid range 328-531 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.