Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXLNB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314509
This item is not returnable.
View return policy
Description
TXLNB Polyclonal antibody specifically detects TXLNB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TXLNB | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
167838 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
taxilin beta | |
Synthetic 17 amino acid peptide from internal region of human TXLNB. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction