Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXLNB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533096
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533096 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533096 Supplier Invitrogen™ Supplier No. PA533096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey (94%) Marmoset (89%).

Promotes motor nerve regeneration. May be involved in intracellular vesicle traffic. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXLNB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TXLNB
Gene Accession No. Q8N3L3
Gene Alias 2310001N14Rik; beta-taxilin; C6orf198; dJ522B19.2; hMDP77; LST001; Mdp77; muscle specific protein; muscle-derived protein 77; RGD1307979; RP3-522B19.2; taxilin beta; Txlnb
Gene Symbols TXLNB
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human TXLNB.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 167838
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.