Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXLNG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544156 Supplier Thermo Scientific Supplier No. PA544156
Add to Cart
Add to Cart

TXLNG Polyclonal Antibody

TXLNG Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TXLNG
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TXLNG
Gene Accession No. Q9NUQ3
Gene Alias 4932441K18Rik; CXHXORF15; CXorf15; ELRG; environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; factor inhibiting ATF4-mediated transcription; FIAT; gamma-taxilin; lipopolysaccharide specific response-5 protein; lipopolysaccharide-responsive gene protein; Lipopolysaccharide-specific response protein 5; Lrg; Lrp; LSR5; taxilin gamma; TXLNG; TXLNGX
Gene Symbols TXLNG
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TXLNG (aa 51-100).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55787
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.