Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNDC Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222328AF647
Description
TXNDC Polyclonal antibody specifically detects TXNDC in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TXNDC | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp564E1962, PDIA11, protein disulfide isomerase family A, member 11, thioredoxin domain containing 1, thioredoxin domain-containing, Thioredoxin domain-containing protein 1, thioredoxin-related transmembrane protein 1, TMXTXNDC, Transmembrane Trx-related protein, TXNDC1thioredoxin domain containing | |
The immunogen this antibody was made to, maps to a region between residue 230 to 280 of human Thioredoxin-Related Transmembrane Protein 1 using the numbering given in entry NP_110382.3 (GeneID 81542). | |
0.1 mL | |
Signal Transduction | |
81542 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction