Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNDC Antibody (OTI3H5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274725F
Description
TXNDC Monoclonal specifically detects TXNDC in Human, Canine, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TXNDC | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TMX1 | |
Full length recombinant protein of human TXNDC (NP_110382) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3H5 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp564E1962, PDIA11, protein disulfide isomerase family A, member 11, thioredoxin domain containing 1, thioredoxin domain-containing, Thioredoxin domain-containing protein 1, thioredoxin-related transmembrane protein 1, TMXTXNDC, Transmembrane Trx-related protein, TXNDC1thioredoxin domain containing | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
81542.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction