Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXNDC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551148
Description
The antibody detects endogenous levels of total TXNDC11 protein.
May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H(2)O(2) generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H(2)O(2) generation. [UniProt].
Specifications
TXNDC11 | |
Polyclonal | |
Unconjugated | |
TXNDC11 | |
2810408E11Rik; AI427833; EF-hand binding protein 1; EF-hand-binding protein 1; EFP1; RGD1307538; thioredoxin domain containing 11; thioredoxin domain-containing protein 11; Txdc11; Txndc11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51061 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6PKC3 | |
TXNDC11 | |
Fusion protein of human TXNDC11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction