Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXNDC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559546
Description
Immunogen sequence: SRTHNVCELC VNQTSGGMKP SSVSVPQCSF FEMAAALDSF YLKEQTFYHV ASDSIECSNF LTSYSPFSYY TACCRTISRG VSGFIDSEQG VFEAPTVAFS S Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H(2)O(2) generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H(2)O(2) generation. [UniProt].
Specifications
TXNDC11 | |
Polyclonal | |
Unconjugated | |
TXNDC11 | |
2810408E11Rik; AI427833; EF-hand binding protein 1; EF-hand-binding protein 1; EFP1; RGD1307538; thioredoxin domain containing 11; thioredoxin domain-containing protein 11; Txdc11; Txndc11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51061 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PKC3 | |
TXNDC11 | |
Recombinant protein corresponding to Human TXNDC11. Recombinant protein control fragment (Product #RP-98732). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction