Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981368
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1368 Supplier Bioss Supplier No. BS11439R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H2O2generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H2O2 generation.

Specifications

Antigen TXNDC11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TXNDC11
Gene Accession No. Q6PKC3
Gene Alias EF-hand binding protein 1; EF-hand-binding protein 1; EFP1; thioredoxin domain containing 11; Thioredoxin domain-containing protein 11; TXNDC11
Gene Symbols TXNDC11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TXNDC11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51061
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.