Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNDC12 Antibody (SR1258), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322499100UL
Description
TXNDC12 Monoclonal antibody specifically detects TXNDC12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
TXNDC12 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1258 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
AG1, AGR1, anterior gradient homolog 1, EC 1.8.4.2, endoplasmic reticulum protein ERp19, Endoplasmic reticulum resident protein 18, Endoplasmic reticulum resident protein 19, endoplasmic reticulum thioredoxin superfamily member, 18 kDa, ER protein 18, ER protein 19, ERP16, ERp18, ERP18AG1, ERP19, hAG-1, hTLP19, PDIA16, protein disulfide isomerase family A, member 16, thioredoxin domain containing 12 (endoplasmic reticulum), thioredoxin domain-containing protein 12, Thioredoxin-like protein p19, TLP19, TLP19hTLP19, TXNDC12 thioredoxin domain containing 12 (endoplasmic reticulum) | |
A synthesized peptide derived from human TXNDC12 (Uniprot #: O95881) | |
100 μg | |
Signal Transduction | |
51060 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction