Learn More
TXNDC17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 208111AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3103
Only null left
Add to Cart
Description
Description
TXNL5 disulfide reductase. May participate in various redox reactions through the reversible oxidation of its active center dithiol to a disulfide and catalyze dithiol-disulfide exchange reactions. Modulates TNF-alpha signaling and NF-kappa-B activation. Has peroxidase activity and may contribute to the elimination of cellular hydrogen peroxide (By similarity).
Specifications
Specifications
TXNDC17
Polyclonal
Unconjugated
TXNDC17
Protein 42 9 9, Thioredoxin like protein 5, TRP14, TXNDC17, TXNL5
Rabbit
Antigen Affinity Chromatography
RUO
287474, 52700, 84817
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BRA2, Q9CQM5
Txndc17
TXNDC17 Fusion Protein Ag14766
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.