Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571321 Supplier Invitrogen™ Supplier No. PA571321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TXNDC2 Polyclonal Antibody for Western Blot

TXNDC2 gene ontology annotations related to this gene include cell redox homeostasis; glycerol ether metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TXNDC2
Gene Accession No. Q86VQ3
Gene Alias spermatid-specific thioredoxin-1; sperm-specific thioredoxin 1; SPTRX; SPTRX1; Sptrx-1; testicular tissue protein Li 215; thioredoxin domain containing 2; thioredoxin domain containing 2 (spermatozoa); thioredoxin domain-containing 2 (spermatozoa); thioredoxin domain-containing protein 2; TXNDC2
Gene Symbols TXNDC2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human TXNDC2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84203
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.