Learn More
Invitrogen™ TXNDC2 Polyclonal Antibody

Description
Immunogen sequence: KELGMESVKA GASGKPEMRL GTQEETSEGD ANESSLLVLS SNVPLLALEF LEIAQAKEKA FLPMVSHTFH MRTEESDASQ EGDDLPKSSA NTSHPKQDDS PKSSEETIQP Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 34%.
Specifications
Specifications
| Antigen | TXNDC2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TXNDC2 |
| Gene Accession No. | Q86VQ3 |
| Gene Alias | spermatid-specific thioredoxin-1; sperm-specific thioredoxin 1; SPTRX; SPTRX1; Sptrx-1; testicular tissue protein Li 215; thioredoxin domain containing 2; thioredoxin domain containing 2 (spermatozoa); thioredoxin domain-containing 2 (spermatozoa); thioredoxin domain-containing protein 2; TXNDC2 |
| Gene Symbols | TXNDC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.