Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXNDC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553967
Description
Immunogen sequence: AQLCDIEEDA ANVAKFMDAF FPDFKKMKSM KLEKTLALLR PNLFHERKDD VLRIIKDEDF KILEQRQVVL SEKEAQALCK EYENED Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.
This gene encodes a protein with an N-terminal thioredoxin domain and three C-terminal nucleoside diphosphate kinase (NDK) domains, but the NDK domains are thought to be catalytically inactive. The sea urchin ortholog of this gene encodes a component of sperm outer dynein arms, and the protein is implicated in ciliary function. Mutations in this gene are implicated in primary ciliary dyskinesia type 6.
Specifications
TXNDC3 | |
Polyclonal | |
Unconjugated | |
NME8 | |
1700056P15Rik; CILD6; epididymis secretory protein Li 99; HEL-S-99; NM23-H8; NME/NM23 family member 8; Nme8; spermatid-specific thioredoxin-2; spermatid-specific thioredoxin-2 protein; sperm-specific thioredoxin 2; SPTRX2; sptrx-2; thioredoxin domain containing 3 (spermatozoa); thioredoxin domain-containing protein 3; Txndc3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N427 | |
NME8 | |
Recombinant protein corresponding to Human TXNDC3. Recombinant protein control fragment (Product #RP-92279). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction