Learn More
Invitrogen™ TXNDC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120532
Description
Positive test controls include: U-87MG, SH-SY5Y, Jurkat. Immunogen sequence: FMDAFFPDFK KMKSMKLEKT LALLRPNLFH ERKDDVLRII KDEDFKILEQ RQVVLSEKEA QALCKEYENE DYFNKLIENM TSGPSLALVL LRDNGLQYWK QLLGPRTVEE AIEYFPESLC AQFAMDSLPV NQLYGSDSLE TAEREIQHFF PLQSTLGLIK PHATSEQREQ ILKIVKEAGF DLTQVKKMFL TPEQIEKIYP KVTGKDFYKD LLEMLSVGPS MVMILTKWNA VAEWRRLMGP TDPEEAKLLS PDSIRAQFGI SKLKNIVHGA SNAYEAKEVV NRLFEDPEEN.
This gene encodes a protein with an N-terminal thioredoxin domain and three C-terminal nucleoside diphosphate kinase (NDK) domains, but the NDK domains are thought to be catalytically inactive. The sea urchin ortholog of this gene encodes a component of sperm outer dynein arms, and the protein is implicated in ciliary function. Mutations in this gene are implicated in primary ciliary dyskinesia type 6.
Specifications
| TXNDC3 | |
| Polyclonal | |
| Unconjugated | |
| NME8 | |
| 1700056P15Rik; CILD6; epididymis secretory protein Li 99; HEL-S-99; NM23-H8; NME/NM23 family member 8; Nme8; spermatid-specific thioredoxin-2; spermatid-specific thioredoxin-2 protein; sperm-specific thioredoxin 2; SPTRX2; sptrx-2; thioredoxin domain containing 3 (spermatozoa); thioredoxin domain-containing protein 3; Txndc3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51314, 73412 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.59 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q715T0, Q8N427 | |
| NME8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 299-588 of human NME8 (NP_057700.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.