Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNDC5 Antibody (OTI2E6), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274726FR
Description
TXNDC5 Monoclonal specifically detects TXNDC5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TXNDC5 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
TXNDC5 | |
Full length recombinant protein of human TXNDC5 (NP_071368) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2E6 | |
Western Blot, Immunohistochemistry | |
ENDOPDI, ERP46, FLJ90810, HCC-2, member 15, MGC3178, PDIA15, thioredoxin domain containing 5, thioredoxin domain containing 5 (endoplasmic reticulum), thioredoxin domain-containing protein 5, thioredoxin related protein, TLP46 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
81567.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction