Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC5 Monoclonal Antibody (OTI1C3), TrueMAB™, OriGene
SDP

Catalog No. 501677037
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7037 Supplier OriGene Supplier No. TA507262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen TXNDC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TXNDC5
Gene Accession No. Q8NBS9, Q91W90
Gene Alias ENDOPDI, ERP46, HCC-2, HCC2, PDIA15, STRF8, UNQ364
Gene Symbols TXNDC5
Host Species Mouse
Immunogen Full length human recombit protein of human TXNDC5 produced in HEK293T cell
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105245, 81567
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.