Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNDC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142727
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142727 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142727 Supplier Invitrogen™ Supplier No. PA5142727
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_110437.2; NP_001139021.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Txndc5
Gene Accession No. Q8NBS9, Q91W90
Gene Alias AL022641; ENDOPDI; endoplasmic reticulum protein ERp46; endoplasmic reticulum resident protein 46; endothelial protein disulphide isomerase; ER protein 46; ERp46; FLJ21353; FLJ90810; HCC2; HCC-2; MGC3178; PC-TRP; PDIA15; Plasma cell-specific thioredoxin-related protein; protein disulfide isomerase family A, member 15; STRF8; thiore; thioredoxin domain containing 5; thioredoxin domain containing 5 (endoplasmic reticulum); thioredoxin domain-containing protein 5; thioredoxin related protein; thioredoxin-like protein p46; Tlp46; TXNDC5; UNQ364; UNQ364/PRO700
Gene Symbols Txndc5
Host Species Goat
Immunogen Peptide with sequence C-RDLESLREYVESQLQ (aa 281-295).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105245, 81567
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.