Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNDC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583686 Supplier Invitrogen™ Supplier No. PA583686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNVARREQPE SLRAQYGTEM PFNAVHGSRD REDADRELAL LFPSLKFSDK DTEAPQGGEA EATAGPTEAL CFPEDVD Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 77%.

May be a regulator of microtubule physiology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NME9
Gene Accession No. Q86XW9
Gene Alias MGC129586; NM23-H9; NME gene family member 9; NME/NM23 family member 9; NME9; Thioredoxin domain-containing protein 6; thioredoxin-like 2; thioredoxin-like protein 2; TXL2; Txl-2; TXNDC6
Gene Symbols NME9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NME9. Recombinant protein control fragment (Product #RP-103981).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.