Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXNDC8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144135
Description
TXNDC8 Polyclonal Antibody for IHC (P), ELISA
TXNDC8 gene ontology annotations related to this gene include cell redox homeostasis; glycerol ether metabolic process.
Specifications
TXNDC8 | |
Polyclonal | |
Unconjugated | |
TXNDC8 | |
bA427L11.2; spermatid-specific thioredoxin-3; spermatocyte/spermatid-specific thioredoxin-3; sperm-specific thioredoxin 3; SPTRX3; SPTRX-3; thioredoxin 6; thioredoxin domain containing 8; thioredoxin domain containing 8 (spermatozoa); thioredoxin domain-containing protein 8; Thioredoxin-6; TRX6; TXNDC8 | |
Rabbit | |
Protein G | |
RUO | |
255220 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q6A555 | |
TXNDC8 | |
Recombinant Human Thioredoxin domain-containing protein 8 protein (1-115AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction