Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TXNDC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556877 Supplier Invitrogen™ Supplier No. PA556877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EADASVDMFS KVLEHQLLQT TKLVEEHLDS EIQKLDQMDE DELERLKEKR LQALRKAQQQ KQEWLSKGHG Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

Not known. associated with cell differentiation. The protein encoded by this gene is a member of the thioredoxin family. The exact function of this protein is not known but it is associated with cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNDC9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TXNDC9
Gene Accession No. O14530
Gene Alias AI098020; APACD; ATP binding protein; ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; ES cell-related protein; fb15e07; PHLP3; Protein 1-4; thioredoxin domain containing 9; thioredoxin domain-containing protein 9; TXNDC9; wu:fb15e07; zgc:55522
Gene Symbols TXNDC9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TXNDC9. Recombinant protein control fragment (Product #RP-96024).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10190
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.