Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TXNIP, Mouse, Unlabeled, Clone: 3A7.1,

Catalog No. MABC605MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC605MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC605MI Supplier MilliporeSigma™ Supplier No. MABC605
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thioredoxin-interacting protein (UniProt Q9H3M7; also known as Thioredoxin-binding protein 2, Vitamin D3 up-regulated protein 1) is encoded by the TXNIP (also known as VDUP1) gene (Gene ID 10628) in human. Thioredoxin interacting protein (TXNIP) binds thioredoxin active cysteine residue and suppresses its antioxidant activity. TXNIP overexpression results in increased cellular ROS level and oxidative cell death, while TXNIP suppression enhances cellular resistance to oxidative injury. TXNIP is also shown to inhibit cellular glucose uptake by downregulating glucose transporter 1 (GLUT1) plasma membrane localization, and TXNIP expression is found to be markedly suppressed in many cancers, making TXNIP a potential anticancer therapeutic target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TXNIP
Applications Western Blot
Classification Monoclonal
Clone 3A7.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene TXNIP, VDUP1
Gene Accession No. Q9H3M7
Host Species Mouse
Immunogen GST-tagged recombinant fragment from the N-terminal half of human TXNIP.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_006463
Test Specificity Clone 3A7.1 targets an epitope within the N-terminal half of TXNIP. Cross-reactivity to human and mouse spliced isoform 2 is likely, but has not been confirmed.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.