Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXNIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549279
Description
Predicted to react with rat based on sequence homology.
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.
Specifications
TXNIP | |
Polyclonal | |
Unconjugated | |
TXNIP | |
1200008J08Rik; AA682105; ARRDC6; EST01027; HHCPA78; hyperlipidemia 1; Hyplip1; Tbp-2; THIF; thioredoxin binding protein 2; thioredoxin binding protein-2; thioredoxin interacting protein; thioredoxin-binding protein 2; Thioredoxin-interacting protein; TXNIP; upregulated by 1,25-dihydroxyvitamin D-3; VDUP1; Vitamin D3 up-regulated protein 1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
56338 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8BG60 | |
TXNIP | |
KLH conjugated synthetic peptide between 116-145 amino acids from the Central region of mouse Txnip. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction