Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNIP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985735
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5735 Supplier Bioss Supplier No. BS3897R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.

Specifications

Antigen TXNIP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TXNIP
Gene Accession No. Q9H3M7
Gene Alias 1200008J08Rik; AA682105; ARRDC6; BOS_3037; EST01027; HHCPA78; hyperlipidemia 1; Hyplip1; Tbp-2; THIF; thioredoxin binding protein 2; thioredoxin binding protein-2; thioredoxin interacting protein; Thioredoxin-binding protein 2; Thioredoxin-interacting protein; Txnip; upregulated by 1,25-dihydroxyvitamin D-3; Vdup1; Vitamin D3 up-regulated protein 1
Gene Symbols TXNIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TXNIP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10628
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.